Learn More
MSC Nilfisk Accessory Kit, 8 pc, 1 1/4 in Hose Diameter, for IVT1000CR
Catalog No. 502839098
Encompass
The Nilfisk Critical Area Vacuum Accessory Kit, L01760480, is an 8-piece set designed to enhance the functionality of IVT1000 and IVT1000CR series vacuums. This comprehensive kit provides essential attachments for effective cleaning in controlled environments, featuring a compatible hose diameter of 1 1/4 inches to ensure efficient debris collection and management.
  • Includes two wands
  • Includes a cap plug
  • Includes a crevice nozzle
  • Includes a dust brush
  • Includes a floor nozzle
  • Includes a hose cuff
  • Includes a stainless steel curved tube
  • Designed for 1 1/4 in hose diameter
  • Compatible with IVT1000CR Safe-Pak and IVT1000CR vacuum models
  • Suitable for dry-only applications
  • Not ESD safe
Catalog No. 50-283-9098 Supplier MSC Supplier No. 35210749
The Nilfisk Critical Area Vacuum Accessory Kit, L01760480, is an 8-piece set designed to enhance the functionality of IVT1000 and IVT1000CR series vacuums. This comprehensive kit provides essential attachments for effective cleaning in controlled environments, featuring a compatible hose diameter of 1 1/4 inches to ensure efficient debris collection and management.
  • Includes two wands
  • Includes a cap plug
  • Includes a crevice nozzle
  • Includes a dust brush
  • Includes a floor nozzle
  • Includes a hose cuff
  • Includes a stainless steel curved tube
  • Designed for 1 1/4 in hose diameter
  • Compatible with IVT1000CR Safe-Pak and IVT1000CR vacuum models
  • Suitable for dry-only applications
  • Not ESD safe