Learn More
Alkali Scientific Vac Check Broth 10 % 10mL, 10mL Vial, 10/Pk
SDP
Catalog No. 5000663243
Encompass_Preferred
Vac Check Broth 10% is a nutrient rich culture medium formulated for sterility testing and microbial contamination detection in vacuum sealed or closed system environments. The 10% formulation supports growth of a wide range of aerobic and facultative microorganisms, making it useful for verifying aseptic processing and packaging integrity. Each sterile vial contains 10mL of prepared broth suitable for convenient sampling and inoculation during laboratory testing procedures. The medium enables reliable detection of microbial contamination in pharmaceutical, medical device, and industrial quality control environments. Supplied in ready to use vials, Vac Check Broth is commonly applied in sterility assurance programs and container validation studies.
Catalog No. 50-006-63243 Supplier Alkali Scientific Supplier No. VCBV00
Vac Check Broth 10% is a nutrient rich culture medium formulated for sterility testing and microbial contamination detection in vacuum sealed or closed system environments. The 10% formulation supports growth of a wide range of aerobic and facultative microorganisms, making it useful for verifying aseptic processing and packaging integrity. Each sterile vial contains 10mL of prepared broth suitable for convenient sampling and inoculation during laboratory testing procedures. The medium enables reliable detection of microbial contamination in pharmaceutical, medical device, and industrial quality control environments. Supplied in ready to use vials, Vac Check Broth is commonly applied in sterility assurance programs and container validation studies.