Learn More
Med Vet International Nobivac Vaccine Intra-Trac KC, 25x1 Dose Tray
Catalog No. 503045264
Encompass_Preferred
An intranasal vaccine recommended for use in healthy susceptible puppies and dogs for prevention of canine infectious tracheobronchitis (“canine cough”) due to canine parainfluenza and B. bronchiseptica.Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet can only ship cold items Monday-Wednesday. *Next Day Air Available With Additional Charges*
Catalog No. 50-304-5264 Supplier Med Vet International Supplier No. PHVINTRATRKC
An intranasal vaccine recommended for use in healthy susceptible puppies and dogs for prevention of canine infectious tracheobronchitis (“canine cough”) due to canine parainfluenza and B. bronchiseptica.Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet can only ship cold items Monday-Wednesday. *Next Day Air Available With Additional Charges*