Learn More
Chemglass Life Sciences Adapter, Vacuum Filtration, #5 or 6 Top Pluro Stopper, 29/42 Lower Inner, Fits Funnel Capacity 350 to 3000mL
SDP
Catalog No. CG105204
Encompass_Preferred
Adapter, Vacuum Filtration, #5 or 6 Top Pluro Stopper, 29/42 Lower Inner, Fits Funnel Capacity 350 to 3000mL. Adapters are used for reduced pressure filtration with plain stem Buchner funnels. Top is tooled to accept the listed size of pluro-stopper. Serrated hose connection has an O.D. of 10mm at the largest serration. FOR PLURO STOPPERS SEE CG-1401.
Catalog No. CG105204 Supplier Chemglass Life Sciences Supplier No. CG105204
Adapter, Vacuum Filtration, #5 or 6 Top Pluro Stopper, 29/42 Lower Inner, Fits Funnel Capacity 350 to 3000mL. Adapters are used for reduced pressure filtration with plain stem Buchner funnels. Top is tooled to accept the listed size of pluro-stopper. Serrated hose connection has an O.D. of 10mm at the largest serration. FOR PLURO STOPPERS SEE CG-1401.