Learn More
MSC Nilfisk Accessory Kit, Aluminum, for Attix Series, 1.375 in Hose
Catalog No. 502838387
Encompass
This accessory kit is designed for use with Attix series vacuums. It features durable aluminum construction and includes three pieces, making it suitable for various industrial cleaning applications.
  • Constructed from durable aluminum
  • Compatible with Attix series vacuums
  • Compatible with 1.375 in diameter hoses
  • Supplied as a 3 piece kit
  • Not ESD safe
Catalog No. 50-283-8387 Supplier MSC Supplier No. 35210756
This accessory kit is designed for use with Attix series vacuums. It features durable aluminum construction and includes three pieces, making it suitable for various industrial cleaning applications.
  • Constructed from durable aluminum
  • Compatible with Attix series vacuums
  • Compatible with 1.375 in diameter hoses
  • Supplied as a 3 piece kit
  • Not ESD safe