Learn More
Chemglass Life Sciences Distilling Column, 24/40 Joint, 250mm, Vacuum Jacketed
SDP
Catalog No. CG122907
Encompass_Preferred
Distilling Column, 24/40 Joint, 250mm, Vacuum Jacketed. Hemple style distilling column, vacuum jacketed to improve performance. Indentations on the inner tube support the packing material. All sizes have an I.D. of approximately 10mm.
Catalog No. CG122907 Supplier Chemglass Life Sciences Supplier No. CG122907
Distilling Column, 24/40 Joint, 250mm, Vacuum Jacketed. Hemple style distilling column, vacuum jacketed to improve performance. Indentations on the inner tube support the packing material. All sizes have an I.D. of approximately 10mm.