Learn More
Genevac Inc Edwards nXDS6i Dry Scroll Pump, Oil-Free Operation, Low Noise Level, 6.2 m³/hr Pumping Speed
Catalog No. NC3432466
Encompass
The Edwards nXDS6i Dry Scroll Vacuum Pump is engineered for clean vacuum applications in scientific environments. Its oil-free design ensures reliability and minimizes contamination, featuring superior vapor handling capabilities and lower operating noise. Benefits include enhanced pumping speed of 6.2 m³/hr, reduced energy usage, and long service intervals of up to 5 years. Designed to support both manual and remote operation, it is ideal for laboratories requiring efficient vacuum solutions.
Catalog No. NC3432466 Supplier Genevac Inc Supplier No. VPNXDS6I
Add to Cart
Add to Cart
Request a Quote
The Edwards nXDS6i Dry Scroll Vacuum Pump is engineered for clean vacuum applications in scientific environments. Its oil-free design ensures reliability and minimizes contamination, featuring superior vapor handling capabilities and lower operating noise. Benefits include enhanced pumping speed of 6.2 m³/hr, reduced energy usage, and long service intervals of up to 5 years. Designed to support both manual and remote operation, it is ideal for laboratories requiring efficient vacuum solutions.