Learn More
MSC ProTeam Vac Station, with One Shelf, for Backpack Vacuums
Catalog No. 502839063
Encompass
This product is a vacuum cleaner repair part, specifically a Vac Station with one shelf, designed to be compatible with backpack vacuums. It is intended for use with various models, including Super Coach Pro, ProVac FS 6, GoFit Corded Series, GoFit Cordless Series, Legacy Backpack Models, and GoFree Flex Pro II.
  • Designed to ensure operators of all heights can back up to the Vac Station and then loop arms through the shoulder straps to begin cleaning.
  • Includes steel wall bracket, mounting rail, screws, and two accessory hooks.
  • Compatible with backpack vacuums.
  • For use with Super Coach Pro, ProVac FS 6, GoFit Corded Series, GoFit Cordless Series, Legacy Backpack Models, and GoFree Flex Pro II.
Catalog No. 50-283-9063 Supplier MSC Supplier No. 40621278
This product is a vacuum cleaner repair part, specifically a Vac Station with one shelf, designed to be compatible with backpack vacuums. It is intended for use with various models, including Super Coach Pro, ProVac FS 6, GoFit Corded Series, GoFit Cordless Series, Legacy Backpack Models, and GoFree Flex Pro II.
  • Designed to ensure operators of all heights can back up to the Vac Station and then loop arms through the shoulder straps to begin cleaning.
  • Includes steel wall bracket, mounting rail, screws, and two accessory hooks.
  • Compatible with backpack vacuums.
  • For use with Super Coach Pro, ProVac FS 6, GoFit Corded Series, GoFit Cordless Series, Legacy Backpack Models, and GoFree Flex Pro II.