Learn More
Ace Glass, Inc. Micro vacuum sublimation apparatus featuring a removable circulatory stem should the researcher prefer to use the head's cold finger with dry ice rather than fluid
SDP
Supplier:  Ace Glass, Inc. 954610
Encompass_Preferred
Micro vacuum sublimation apparatus featuring a removable circulatory stem should the researcher prefer to use the head's cold finger with dry ice rather than fluid
Catalog No. 50-976-997