Learn More
Chemglass Life Sciences Vacuum Trap, Serrated Hose Connections, Complete, Trap Bottle is 32mm Body OD x 200mm Overall Length, 29/42 Jt.
SDP
Catalog No. CG451501
Encompass_Preferred
Vacuum Trap, Serrated Hose Connections, Complete, Trap Bottle is 32mm Body OD x 200mm Overall Length, 29/42 Jt. Two piece construction for easy removal of condensate. Inlet and outlet are 180º apart for installation into a variety of vacuum systems. Stopper has a standard taper inner joint and body has an outer joint. Body length is the overall length from the top of the joint to the bottom of the round bottom. Serrated hose connections have an O.D. of 13mm at the smallest serration and 16mm at the largest serration.
Catalog No. CG451501 Supplier Chemglass Life Sciences Supplier No. CG451501
Vacuum Trap, Serrated Hose Connections, Complete, Trap Bottle is 32mm Body OD x 200mm Overall Length, 29/42 Jt. Two piece construction for easy removal of condensate. Inlet and outlet are 180º apart for installation into a variety of vacuum systems. Stopper has a standard taper inner joint and body has an outer joint. Body length is the overall length from the top of the joint to the bottom of the round bottom. Serrated hose connections have an O.D. of 13mm at the smallest serration and 16mm at the largest serration.