Learn More
Chemglass Life Sciences Vacuum Trap, Complete, 32mm Body OD x 200mm Overall Length, 10mm OD Side and Inner Tubes, 29/42 Jt.
SDP
Catalog No. CG451601
Encompass_Preferred
Vacuum Trap, Complete, 32mm Body OD x 200mm Overall Length, 10mm OD Side and Inner Tubes, 29/42 Jt. Similar to CG-4514, but with the top inlet tube bent at a 90º angle.
Catalog No. CG451601 Supplier Chemglass Life Sciences Supplier No. CG451601
Vacuum Trap, Complete, 32mm Body OD x 200mm Overall Length, 10mm OD Side and Inner Tubes, 29/42 Jt. Similar to CG-4514, but with the top inlet tube bent at a 90º angle.