Learn More
Chemglass Life Sciences Vacuum Trap, Solvent Vapor, Plastic Coated, Chem-Drain Valve, Complete
SDP
Catalog No. CG452301
Encompass_Preferred
Vacuum Trap, Solvent Vapor, Plastic Coated, Chem-Drain Valve, Complete. Similar to CG-4519, but modified with a 6mm Chem-Drain valve for use under high vacuum applications. Waste solvent drains directly through the PTFE plug, eliminating fragile drain tubes. Dimensions are the same as CG-4519.
Catalog No. CG452301 Supplier Chemglass Life Sciences Supplier No. CG452301
Vacuum Trap, Solvent Vapor, Plastic Coated, Chem-Drain Valve, Complete. Similar to CG-4519, but modified with a 6mm Chem-Drain valve for use under high vacuum applications. Waste solvent drains directly through the PTFE plug, eliminating fragile drain tubes. Dimensions are the same as CG-4519.