Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Tygon E-3603, Non-DEHP Vacuum, 1/4"" ID x 5/8"" OD; 10 ft
Catalog No. NC1631020
Encompass
Applications: General lab, food and beverage, and biopharm. Vacuum service of 29.9 Hg (760 mm Hg) at 73F (23C). Characteristics: Non-DEHP, phthalate-free plasticizer. Flexible, biodegradable, crack-resistant, nontoxic, and nonoxidizing. Clear.
Catalog No. NC1631020 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0641420
Applications: General lab, food and beverage, and biopharm. Vacuum service of 29.9 Hg (760 mm Hg) at 73F (23C). Characteristics: Non-DEHP, phthalate-free plasticizer. Flexible, biodegradable, crack-resistant, nontoxic, and nonoxidizing. Clear.