Learn More
Accuflow Systems Saint-Gobain Vacuum Tubing, Tygon S3, 1/4 in ID, 50 ft
Catalog No. 502365760
Encompass_Preferred
Tygon S3 E-3603 vacuum tubing is a non-DEHP laboratory-grade tubing formulated for vacuum and instrumentation applications. It features a smooth inner bore to minimize buildup, resistance to flex-fatigue and abrasion, and a bio-based formulation without intentionally added phthalates or BPA. The tubing is suitable for full vacuum at room temperature and is supplied in 50 ft (15 m) coils.
  • Non-DEHP laboratory tubing suitable for vacuum use.
  • Smooth inner bore for reduced buildup and easy cleaning.
  • Resists flex-fatigue and abrasion for extended service life.
  • Available in 1/4 in (6.4 mm) ID, 5/8 in (15.9 mm) OD, 3/16 in (4.8 mm) wall.
  • Supplied as 50 ft (15 m) coils for instrumentation and vacuum lines.
Catalog No. 50-236-5760 Supplier Accuflow Systems Supplier No. ACF00020
Tygon S3 E-3603 vacuum tubing is a non-DEHP laboratory-grade tubing formulated for vacuum and instrumentation applications. It features a smooth inner bore to minimize buildup, resistance to flex-fatigue and abrasion, and a bio-based formulation without intentionally added phthalates or BPA. The tubing is suitable for full vacuum at room temperature and is supplied in 50 ft (15 m) coils.
  • Non-DEHP laboratory tubing suitable for vacuum use.
  • Smooth inner bore for reduced buildup and easy cleaning.
  • Resists flex-fatigue and abrasion for extended service life.
  • Available in 1/4 in (6.4 mm) ID, 5/8 in (15.9 mm) OD, 3/16 in (4.8 mm) wall.
  • Supplied as 50 ft (15 m) coils for instrumentation and vacuum lines.