Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Tygon E-3603, Non-DEHP Vacuum, 5/8"" ID x 1 3/8"" OD; 10 ft
Catalog No. NC1675494
Encompass
Formulation: Tygon E-3603 vacuum Applications: General lab, food and beverage, and biopharm. Vacuum service of 29.9" Hg (760 mm Hg) at 73¿F (23¿C). Characteristics: Non-DEHP, phthalate-free plasticizer. Flexible, biodegradable, crack-resistant, nontoxic, and nonoxidizing. Clear. Certifications: USP Class VI and NSF 51 Temperature range: Brittle temperature is -51F (-46¿C). Sterilize: Ethylene oxide or autoclave for 30 min at 250¿F (121¿C), 15 psi (1 bar).
Catalog No. NC1675494 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0641455
Formulation: Tygon E-3603 vacuum Applications: General lab, food and beverage, and biopharm. Vacuum service of 29.9" Hg (760 mm Hg) at 73¿F (23¿C). Characteristics: Non-DEHP, phthalate-free plasticizer. Flexible, biodegradable, crack-resistant, nontoxic, and nonoxidizing. Clear. Certifications: USP Class VI and NSF 51 Temperature range: Brittle temperature is -51F (-46¿C). Sterilize: Ethylene oxide or autoclave for 30 min at 250¿F (121¿C), 15 psi (1 bar).