Learn More
Accuflow Systems Tygon Tubing, Thermoplastic, 5/8 in I.D., 50 ft
Catalog No. 502365786
Encompass_Preferred
Clear thermoplastic tubing designed for vacuum and general fluid transfer applications. The tubing provides visual clarity for inspection, flexibility for routing, and sufficient working pressure and vacuum performance for laboratory use.
  • Thermoplastic construction for flexibility and chemical compatibility.
  • 5/8 in inner diameter, 3/8 in wall thickness, 50 ft coil length.
  • Crystal-clear appearance for visual monitoring of fluids.
  • Maximum working pressure 43 psi at 73°F and recommended maximum temperature 165°F (74°C).
  • Maintains vacuum integrity suitable for typical laboratory vacuum service.
Catalog No. 50-236-5786 Supplier Accuflow Systems Supplier No. ACF00049
Clear thermoplastic tubing designed for vacuum and general fluid transfer applications. The tubing provides visual clarity for inspection, flexibility for routing, and sufficient working pressure and vacuum performance for laboratory use.
  • Thermoplastic construction for flexibility and chemical compatibility.
  • 5/8 in inner diameter, 3/8 in wall thickness, 50 ft coil length.
  • Crystal-clear appearance for visual monitoring of fluids.
  • Maximum working pressure 43 psi at 73°F and recommended maximum temperature 165°F (74°C).
  • Maintains vacuum integrity suitable for typical laboratory vacuum service.