Learn More
Chemglass Life Sciences Chemglass Heavy Wall Vacuum Tubing (Excelon), PVC, 3/8 in ID x 7/8 in OD x 10 ft (CG-735-V-375)
SDP
Catalog No. NC2323688
Encompass_Preferred
Chemglass heavy wall vacuum tubing is a laboratory-grade PVC tubing designed for vacuum and general fluid transfer applications. The tubing features a thick wall for increased durability and kink resistance, and is compatible with autoclaving and gamma sterilization.
  • Heavy-wall PVC tubing suitable for vacuum use.
  • Inner diameter: 3/8 in; outer diameter: 7/8 in; wall: 1/4 in.
  • Sold as a 10 ft roll for convenient installation.
  • Durable durometer hardness (Shore A 55) for long service life.
  • Temperature range -58°F to 165°F for varied laboratory conditions.
  • Compatible with autoclave and gamma sterilization methods.
  • Meets FDA material standards for appropriate applications.
Catalog No. NC2323688 Supplier Chemglass Life Sciences Supplier No. CG735V375
Chemglass heavy wall vacuum tubing is a laboratory-grade PVC tubing designed for vacuum and general fluid transfer applications. The tubing features a thick wall for increased durability and kink resistance, and is compatible with autoclaving and gamma sterilization.
  • Heavy-wall PVC tubing suitable for vacuum use.
  • Inner diameter: 3/8 in; outer diameter: 7/8 in; wall: 1/4 in.
  • Sold as a 10 ft roll for convenient installation.
  • Durable durometer hardness (Shore A 55) for long service life.
  • Temperature range -58°F to 165°F for varied laboratory conditions.
  • Compatible with autoclave and gamma sterilization methods.
  • Meets FDA material standards for appropriate applications.