Learn More
Aviva Systems Biology Corp VAC14 Antibody - C-terminal region (ARP61721_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61721_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VAC14. It was validated on Western Blot. . Alternate names: ArPIKfyve, FLJ10305, FLJ36622, FLJ46582, MGC149815, MGC149816, TAX1BP2, TRXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-450