Learn More
RIVERDALE SCIENTIFIC LLC VacBuddy Vacuum TRAP aspiration kit, HDPE 2 liter x 2 Liter bottles from Riverdale Scientfic LLC
Catalog No. NC2884355
Encompass_Preferred
VacBuddy follows guidelines laid out by CDC to provide you a complete Vacuum Trap Aspiration Kit. This kit comprises two 2-liter vacuum rated bottles with a 360 degree 'swivel' cap that houses angled barbs to ensure no-kinking of the tubing, thereby enabling unrestricted aspiration of fluids.•Vent filter is a PTFE hydrophobic membrane with a pore-size of 0.22um to protect the vacuum source. •Tubing does not kink due to unique design. •Cap is thick with an O-ring which makes a tight seal and is chemically resistant •Barbs are color coded to avoid confusion to where to attach tubing.•Ample length of tubing is provided to suit one's unique requirements.
Catalog No. NC2884355 Supplier RIVERDALE SCIENTIFIC LLC Supplier No. RSVACBUD2L2
VacBuddy follows guidelines laid out by CDC to provide you a complete Vacuum Trap Aspiration Kit. This kit comprises two 2-liter vacuum rated bottles with a 360 degree 'swivel' cap that houses angled barbs to ensure no-kinking of the tubing, thereby enabling unrestricted aspiration of fluids.•Vent filter is a PTFE hydrophobic membrane with a pore-size of 0.22um to protect the vacuum source. •Tubing does not kink due to unique design. •Cap is thick with an O-ring which makes a tight seal and is chemically resistant •Barbs are color coded to avoid confusion to where to attach tubing.•Ample length of tubing is provided to suit one's unique requirements.