Learn More
RIVERDALE SCIENTIFIC LLC VacBuddy Vacuum TRAP aspiration kit, HDPE 2 liter x 1 Liter bottles from Riverdale Scientfic LLC
Catalog No. NC3728581
Encompass_Preferred
VacBuddy follows guidelines laid out by CDC to provide you a complete Vacuum Trap Aspiration Kit. This kit contains a 2 Liter and 1 Liter vacuum rated bottles with a 360 degree 'swivel' cap that houses angled barbs to ensure no-kinking of the tubing, thereby enabling unrestricted aspiration of fluids.•Vent filter is a PTFE hydrophobic membrane with a pore-size of 0.22um to protect the vacuum source. •Tubing does not kink due to unique cap design. •Cap is thick with an O-ring which makes a tight seal and is chemically resistant •Secondary ray is provided•Ample length of tubing is provided to suit one's unique requirements.
Catalog No. NC3728581 Supplier RIVERDALE SCIENTIFIC LLC Supplier No. RSVACBUD2L1
VacBuddy follows guidelines laid out by CDC to provide you a complete Vacuum Trap Aspiration Kit. This kit contains a 2 Liter and 1 Liter vacuum rated bottles with a 360 degree 'swivel' cap that houses angled barbs to ensure no-kinking of the tubing, thereby enabling unrestricted aspiration of fluids.•Vent filter is a PTFE hydrophobic membrane with a pore-size of 0.22um to protect the vacuum source. •Tubing does not kink due to unique cap design. •Cap is thick with an O-ring which makes a tight seal and is chemically resistant •Secondary ray is provided•Ample length of tubing is provided to suit one's unique requirements.