Learn More
Medchemexpress LLC HY-N1419 5mg Medchemexpress, Vaccarin CAS:53452-16-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN14195MG
Encompass_Preferred
Medchemexpress, HY-N1419 5mg Vaccarin CAS:53452-16-7 Vaccarin is an active flavonoid glycoside associated with various biological functions. Vaccarin significantly promote wound healing and endothelial cells and fibroblasts proliferation in the wound site. Vaccarin ameliorates insulin resistance and steatosis by activating the AMPK signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2844