Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Vaccinated Labels

Catalog No. 19129859 Shop All Brady Corporation Products
Change view
Click to view available options
Mounting Type:
Self-adhesive
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
19-129-859 Self-adhesive
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-859 Supplier Brady™ Supplier No. 152583
  • $111.00
    $91.65 / Pack of 1

    Save $19.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labels let people know who has been vaccinated, help identify individuals quickly, can be used in offices, hospitals, institutions, retail and industrial settings

  • B-747 polyester material with permanent acrylic adhesive
  • Good solvent resistance
  • Performs well in high-temperature areas
  • Accepts pen and permanent marker ink

Specifications

Material Polyester (B-747)
Mounting Type Self-adhesive
Height (English) 0.625 in.
Height (Metric) 1.5875 cm
Width (English) 1.5 in.
Width (Metric) 3.81 cm
Shape Rectangular
Languages English
Displayed Text VACCINATED
Product Type Vaccinated Labels
Color Green on White
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.