Learn More
Med Vet International Nobivac Vaccine 3-RABIES, 50X1 Dose Trays
Catalog No. 503045294
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Rabies Vaccine, Killed Virus, 3-year in dogs and cats, 1-year in cattle and sheep. For use in dogs, cats, cattle and sheep only. Preservative: Gentamicin. Meat withdrawal: 21 days.
Catalog No. 50-304-5294 Supplier Med Vet International Supplier No. RXV3RABIES50D
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Rabies Vaccine, Killed Virus, 3-year in dogs and cats, 1-year in cattle and sheep. For use in dogs, cats, cattle and sheep only. Preservative: Gentamicin. Meat withdrawal: 21 days.