Learn More
Med Vet International NobIVac 3-Rabies Vaccine, 10 Dose Tank
Catalog No. 502828393
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Helps prevent rabies For use in dogs and cats (every 3 years)Cattle and sheep, (every year)Killed virus Injectable vaccine Made in the USA Vaccines are non-returnable Shipping Refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. Next day air available for additional charge.
Catalog No. 50-282-8393 Supplier Med Vet International Supplier No. RXV3RABIES10D
 Restricted Item
Add to Cart
Add to Cart
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Helps prevent rabies For use in dogs and cats (every 3 years)Cattle and sheep, (every year)Killed virus Injectable vaccine Made in the USA Vaccines are non-returnable Shipping Refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. Next day air available for additional charge.