Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Vaccine Vial Gripper
GREENER_CHOICE

Catalog No. 17100482 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Assorted (Red, Blue, Green, Yellow, Purple)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-100-482 Assorted (Red, Blue, Green, Yellow, Purple)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-482
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The perfect Vaccine grip for optimal precision

  • These two styles of hand-operated Grippers are designed to pick up cryogenic and crimped 10R-size vaccine/serum vials, respectively
  • Both are durably made of reinforced polypropylene which is cold-tolerant and resistant to everyday wear
  • The designated squeeze target ensures maximum precision and grip with every use
  • Each style is available in a pack of 5 assorted colors for easy identification and organization
  • A combo pack with one of each style is also available

Specifications

Type Vaccine Gripper
Color Assorted (Red, Blue, Green, Yellow, Purple)
For Use With (Equipment) Vaccine Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.