Learn More
Bon Opus Biosciences Recombinant Vacciniavirus B18R 500ug
SDP
Supplier:  Bon Opus Biosciences CM32500UG
Encompass_Preferred
Recombinant Vacciniavirus B18R is produced by our Mammalian expression system and the target gene encoding His20-Glu351 is expressed with a 6His tag at the C-terminus. Accession# P25213. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH7.4. Bon Opus Cat# CM32
Catalog No. 50-217-2821