Learn More
ATCC Vaccinia virus; Strain: vTF7-3 [Wr]
Catalog No. 502384147
Encompass
Vaccinia virus strain vTF7-3 [Wr] is propagated in BS-C-1 cells (ATCC CCL-26). This strain was constructed in vitro by transfecting vaccinia virus-infected cells with pTF703 cells (ATCC 67202) and was deposited by the National Institutes of Health. It expresses bacteriophage T[7] RNA polymerase. This product requires a VS 16-6A permit, which can be downloaded at the USDA website here. Please submit the completed permit form to SalesPermits@atcc.org with your ATCC sales order number and/or your purchase number noted in the subject line and body of the email.
Catalog No. 50-238-4147 Supplier ATCC Supplier No. VR2153
 Restricted Item
Add to Cart
Add to Cart
Vaccinia virus strain vTF7-3 [Wr] is propagated in BS-C-1 cells (ATCC CCL-26). This strain was constructed in vitro by transfecting vaccinia virus-infected cells with pTF703 cells (ATCC 67202) and was deposited by the National Institutes of Health. It expresses bacteriophage T[7] RNA polymerase. This product requires a VS 16-6A permit, which can be downloaded at the USDA website here. Please submit the completed permit form to SalesPermits@atcc.org with your ATCC sales order number and/or your purchase number noted in the subject line and body of the email.