Learn More
Sino Biological Vaccinia virus (VACV) (strain Copenhagen) A33R Antibody, Rabbit PAb, Antigen Affinity Purified 20 µL
Catalog No. 5000748811
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Vaccinia virus (VACV) (strain Copenhagen) A33R ( Catalog#40896-V07E; P68616; Val57-Asn185). A33R Protein specific IgG was purified by Vaccinia virus (VACV) (strain Copenhagen) A33R affinity chromatography.
Catalog No. 50-007-48811 Supplier Sino Biological Supplier No. 40896T3020
Produced in rabbits immunized with purified, recombinant Vaccinia virus (VACV) (strain Copenhagen) A33R ( Catalog#40896-V07E; P68616; Val57-Asn185). A33R Protein specific IgG was purified by Vaccinia virus (VACV) (strain Copenhagen) A33R affinity chromatography.