Learn More
Cayman Chemical Primin, Purity: ≥98%, CAS: 15121-94-5, 10 mg
SDP
Catalog No. 502913996
Encompass_Preferred
A benzoquinone with diverse biological activities; selectively inhibits COX-2 over COX-1 (IC50s = 0.09 and 5.14 µM, respectively); induces the production of adrenochrome from epinephrine in a cell-free assay at 8.81 µM; active against T. brucei rhodesiense and L. donovani (IC50s = 0.144 and 0.711 µM, respectively), as well as S. aureus (IC50 = 8.94 µM); induces contact sensitization in guinea pigs
Catalog No. 50-291-3996 Supplier Cayman Chemical Supplier No. 3544110 MG
A benzoquinone with diverse biological activities; selectively inhibits COX-2 over COX-1 (IC50s = 0.09 and 5.14 µM, respectively); induces the production of adrenochrome from epinephrine in a cell-free assay at 8.81 µM; active against T. brucei rhodesiense and L. donovani (IC50s = 0.144 and 0.711 µM, respectively), as well as S. aureus (IC50 = 8.94 µM); induces contact sensitization in guinea pigs