Learn More
Cole-Parmer Fomblin DC976VF Vacuum Grease, Dimethyle siloxane / Silicon dioxide; 5.3 oz
Supplier:  Cole-Parmer 7975130
Encompass_Preferred
Vacuum grease is a stiff, nonmelting silicone lubricant that maintains its consistency from 40 to 500F. Grease is nonoxidizing and nongumming and has excellent resistance to water and chemicals.
Catalog No. NC9053548