Learn More
Research Products International Corp Vaccupette, 96 Channel Pipettor
SDP
Catalog No. 50998538
Encompass
Aspirate or dispense 96 wells in less than 10 seconds speeding up repetitive washing, dispensing or aspirating of wells by working simultaneously. Connect a disposable plastic syringe and withdraw the plunger to transfer liquid.Made of transparent plastic material, it has a vacuum outlet at one end, and a short length of 3.2 mm inner diameter (1/8 Inch) tubing is supplied for syringe connection. Sterile/Disposable model is molded of clear, disposable plastic that can be used only once and cannot be autoclaved or gas sterilized.
Catalog No. 50-998-538 Supplier Research Products International Corp Supplier No. F378760000
Aspirate or dispense 96 wells in less than 10 seconds speeding up repetitive washing, dispensing or aspirating of wells by working simultaneously. Connect a disposable plastic syringe and withdraw the plunger to transfer liquid.Made of transparent plastic material, it has a vacuum outlet at one end, and a short length of 3.2 mm inner diameter (1/8 Inch) tubing is supplied for syringe connection. Sterile/Disposable model is molded of clear, disposable plastic that can be used only once and cannot be autoclaved or gas sterilized.