Learn More
Bioworld Vacin and Went Medium, 25 L
Catalog No. 50254795
Encompass_Preferred
Vacin & Went MediumContains Micronutrients and Macronutrients as described by Vacin & Went (1949)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 1.67 g/LSoluble in water
Catalog No. 50-254-795 Supplier Bioworld Supplier No. 306301182
Vacin & Went MediumContains Micronutrients and Macronutrients as described by Vacin & Went (1949)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 1.67 g/LSoluble in water