Learn More
Bioworld Vacin & Went Orchid Medium, w/ Agar and Sucrose, 25 L
Catalog No. 50254987
Encompass_Preferred
Vacin & Went Orchid MediumContains the Macronutrients and Micronutrients as described by Vacin & Went (1949) with the addition of Agar and SucroseContains Ferrous Sulfate in place of Ferric TartrateCytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 28.67 g/LPartially soluble in water
Catalog No. 50-254-987 Supplier Bioworld Supplier No. 306301953
Vacin & Went Orchid MediumContains the Macronutrients and Micronutrients as described by Vacin & Went (1949) with the addition of Agar and SucroseContains Ferrous Sulfate in place of Ferric TartrateCytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 28.67 g/LPartially soluble in water