Learn More
Electron Microscopy Sciences Dupont Molykote High Vacuum Grease
SDP
Catalog No. 501898974
Encompass_Preferred
A silicone lubricant for glass-to-glass joints and glass-to-rubber connections. Resistant to most chemicals and is inert and heat stable.
Catalog No. 50-189-8974 Supplier Electron Microscopy Sciences Supplier No. 60705
A silicone lubricant for glass-to-glass joints and glass-to-rubber connections. Resistant to most chemicals and is inert and heat stable.