Learn More
Electron Microscopy Sciences Vacuum Wand Hard Anodized Wafer Tip for 8" (200mm) wafers
SDP
Catalog No. 501931162
Encompass_Preferred
Made from hard anodized aluminum, these wafer tips are able to be used in the Porta-Wand, Porta-Vac II and EMS Vacuum Wand Kits. Withstand high temperatures up to 250°C. They feature a 3" long, straight stainless steel tube covered by a PTFE sleeve. The press-fit adapter is made from static dissapative plastic.
Catalog No. 50-193-1162 Supplier Electron Microscopy Sciences Supplier No. 7193602
Made from hard anodized aluminum, these wafer tips are able to be used in the Porta-Wand, Porta-Vac II and EMS Vacuum Wand Kits. Withstand high temperatures up to 250°C. They feature a 3" long, straight stainless steel tube covered by a PTFE sleeve. The press-fit adapter is made from static dissapative plastic.