Learn More
Electron Microscopy Sciences Vacuum Wand Molded PEEK Tip for 8" (200mm) wafers, ESD-Safe
SDP
Catalog No. 501931160
Encompass_Preferred
Wafer tips made from PEEK, a high performance plastic and are able to be used in Porta-Wand, Porta-Vac II, and EMS Vacuum Wand Kits. Withstand temperatures up to 100°C. Thin profile to allow easy access to wafers.
Catalog No. 50-193-1160 Supplier Electron Microscopy Sciences Supplier No. 7193502
Wafer tips made from PEEK, a high performance plastic and are able to be used in Porta-Wand, Porta-Vac II, and EMS Vacuum Wand Kits. Withstand temperatures up to 100°C. Thin profile to allow easy access to wafers.