Learn More
Medchemexpress LLC Vacquinol-1 dihydrochloride | 2309312-85-2 | 99.7% | 425.78 | 5 MG
SDP
Catalog No. 5000384750
Encompass_Preferred
Vacquinol-1 dihydrochloride (NSC13316 dihydrochloride) is a MKK4-specific activator that activates MAPK pathways. It specifically induces human glioblastoma cell (GC) death, attenuates tumor progression, and prolongs survival in a glioblastoma multiforme (GBM) mouse model. It also induces apoptosis in hepatocellular carcinoma (HCC) cells.
  • MKK4-specific activator
  • Activates MAPK pathways
  • Induces human glioblastoma cell (GC) death
  • Attenuates tumor progression
  • Prolongs survival in glioblastoma multiforme (GBM) mouse model
  • Induces apoptosis in hepatocellular carcinoma (HCC) cells
Catalog No. 50-003-84750 Supplier Medchemexpress LLC Supplier No. HY19345A5MG
Vacquinol-1 dihydrochloride (NSC13316 dihydrochloride) is a MKK4-specific activator that activates MAPK pathways. It specifically induces human glioblastoma cell (GC) death, attenuates tumor progression, and prolongs survival in a glioblastoma multiforme (GBM) mouse model. It also induces apoptosis in hepatocellular carcinoma (HCC) cells.
  • MKK4-specific activator
  • Activates MAPK pathways
  • Induces human glioblastoma cell (GC) death
  • Attenuates tumor progression
  • Prolongs survival in glioblastoma multiforme (GBM) mouse model
  • Induces apoptosis in hepatocellular carcinoma (HCC) cells