Learn More
Medchemexpress LLC Vacquinol-1 dihydrochloride (NSC13316 dihydrochloride) | 2309312-85-2 | 99.7% | 425.78 | 100 MG
SDP
Catalog No. 5000386000
Encompass_Preferred
Vacquinol-1 (NSC13316) dihydrochloride is a MKK4-specific activator that activates MAPK pathways. It specifically induces human glioblastoma cell (GC) death, attenuates tumor progression, and prolongs survival in a glioblastoma multiforme (GBM) mouse model. Vacquinol-1 dihydrochloride also induces apoptosis in hepatocellular carcinoma (HCC) cells.
  • Activates MAPK pathways
  • Induces human glioblastoma cell death
  • Attenuates tumor progression
  • Prolongs survival in a glioblastoma multiforme mouse model
  • Induces apoptosis in hepatocellular carcinoma cells
Catalog No. 50-003-86000 Supplier Medchemexpress LLC Supplier No. HY19345A100MG
Vacquinol-1 (NSC13316) dihydrochloride is a MKK4-specific activator that activates MAPK pathways. It specifically induces human glioblastoma cell (GC) death, attenuates tumor progression, and prolongs survival in a glioblastoma multiforme (GBM) mouse model. Vacquinol-1 dihydrochloride also induces apoptosis in hepatocellular carcinoma (HCC) cells.
  • Activates MAPK pathways
  • Induces human glioblastoma cell death
  • Attenuates tumor progression
  • Prolongs survival in a glioblastoma multiforme mouse model
  • Induces apoptosis in hepatocellular carcinoma cells