Learn More
Medchemexpress LLC Vactosertib Hydrochloride | 1352610-25-3 | 99.2% | 435.88 | 50 MG
SDP
Catalog No. 5000401544
Encompass_Preferred
Vactosertib Hydrochloride is a potent, orally active, and ATP-competitive activin receptor-like kinase 5 (ALK5) inhibitor with an IC50 of 12.9 nM. It also inhibits ALK2 and ALK4 at nanomolar concentrations (IC50 of 17.3 nM). Vactosertib Hydrochloride demonstrates potent antimetastatic activity and anticancer effects. This product is for research use only.
  • Potent and orally active ALK5 inhibitor
  • Inhibits ALK2 and ALK4 at nanomolar concentrations
  • Demonstrates antimetastatic activity
  • Exhibits anticancer effects
  • Intended for research use only
Catalog No. 50-004-01544 Supplier Medchemexpress LLC Supplier No. HY19928A50MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vactosertib Hydrochloride is a potent, orally active, and ATP-competitive activin receptor-like kinase 5 (ALK5) inhibitor with an IC50 of 12.9 nM. It also inhibits ALK2 and ALK4 at nanomolar concentrations (IC50 of 17.3 nM). Vactosertib Hydrochloride demonstrates potent antimetastatic activity and anticancer effects. This product is for research use only.
  • Potent and orally active ALK5 inhibitor
  • Inhibits ALK2 and ALK4 at nanomolar concentrations
  • Demonstrates antimetastatic activity
  • Exhibits anticancer effects
  • Intended for research use only