Learn More
Foxx Life Sciences Vacuum Trap System (Vacuum Flask Alternative) for Aspiration and Vacuum Protection, Shatterproof 4 Liter Aspirator Bottle Assembly with Secondary Container, Bleach Compatible HDPE, 4L Bottle with Inline Vent Filter
GREENER_CHOICE
Catalog No. 501489321
Encompass_Preferred
302-4001-FLS: Vactrap™, HDPE, 4L, Red Bin, ID TubingFoxx Life Sciences Vactrap is the worlds most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap comes ready to usesimply attach to your vacuum and begin your work. The full Vactrap system includes Foxx Heavy Duty Vacuum Bottle, with a patented VersaCap closure for tangle-free openingno need to disconnect tubing, ever. A 0.2m PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nested neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for modulation of vacuum. Other Vactrap options offer one bottle, or setups without secondary containment. Say goodbye to glass. Say hello to Vactrap.
Catalog No. 50-148-9321 Supplier Foxx Life Sciences Supplier No. 3024001FLS
302-4001-FLS: Vactrap™, HDPE, 4L, Red Bin, ID TubingFoxx Life Sciences Vactrap is the worlds most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap comes ready to usesimply attach to your vacuum and begin your work. The full Vactrap system includes Foxx Heavy Duty Vacuum Bottle, with a patented VersaCap closure for tangle-free openingno need to disconnect tubing, ever. A 0.2m PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nested neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for modulation of vacuum. Other Vactrap options offer one bottle, or setups without secondary containment. Say goodbye to glass. Say hello to Vactrap.