Learn More
Foxx Life Sciences VACTRAP ASPIRATOR PP 1 1L BIN
GREENER_CHOICE
Catalog No. 502060936
Encompass_Preferred
Foxx Life Sciences' Vactrap2™ is the world's most advanced vacuum trap system.  Ideal for the protection of vacuum lines and scientists alike, Vactrap2™ safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration.  Best of all, Vactrap2™ comes ready to use; simply attach to your vacuum and begin your work. The full Vactrap2™ system includes two Foxx Heavy Duty Vacuum Bottles (autoclavable PP or chemically-resistant HDPE), each with a patented VersaCap closure for tangle-free opening, no need to disconnect tubing, ever.  NEW Improved heavy-duty EZBio TPE V tubing to avoid kinking.  A 0.2µm PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nestled neatly in a secondary container with a small footprint and incredible stability.  A pinch clamp allows for the modulation of vacuum.  Other Vactrap2™ options offer one bottle or setups without secondary containment. 
Catalog No. 50-206-0936 Supplier Foxx Life Sciences Supplier No. 3051101FLS
Foxx Life Sciences' Vactrap2™ is the world's most advanced vacuum trap system.  Ideal for the protection of vacuum lines and scientists alike, Vactrap2™ safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration.  Best of all, Vactrap2™ comes ready to use; simply attach to your vacuum and begin your work. The full Vactrap2™ system includes two Foxx Heavy Duty Vacuum Bottles (autoclavable PP or chemically-resistant HDPE), each with a patented VersaCap closure for tangle-free opening, no need to disconnect tubing, ever.  NEW Improved heavy-duty EZBio TPE V tubing to avoid kinking.  A 0.2µm PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nestled neatly in a secondary container with a small footprint and incredible stability.  A pinch clamp allows for the modulation of vacuum.  Other Vactrap2™ options offer one bottle or setups without secondary containment. 