Learn More
Foxx Life Sciences Vactrap™ Vacuum Trap System (Vacuum Flask Alternative) for Aspiration and Vacuum Protection, Shatterproof 2 Liter Aspirator Bottle Assembly with Secondary Container, Autoclavable Polypropylene (PP), 2L Bottle with Inline Vent Filter
GREENER_CHOICE
Catalog No. 501489322
Encompass_Preferred
305-2001-FLS: Vactrap™, PP, 2L, Red Bin, ID TubingFoxx Life Sciences Vactrap is the worlds most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap comes ready to usesimply attach to your vacuum and begin your work. The full Vactrap system includes one Foxx Heavy Duty Vacuum Bottle (autoclavable PP or chemically-resistant HDPE), each with a patented VersaCap closure for tangle-free openingno need to disconnect tubing, ever. A 0.2m PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nested neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for modulation of vacuum. Other Vactrap options offer one bottle, or setups without secondary containment. Say goodbye to glass. Say hello to Vactrap.
Catalog No. 50-148-9322 Supplier Foxx Life Sciences Supplier No. 3052001FLS
305-2001-FLS: Vactrap™, PP, 2L, Red Bin, ID TubingFoxx Life Sciences Vactrap is the worlds most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap comes ready to usesimply attach to your vacuum and begin your work. The full Vactrap system includes one Foxx Heavy Duty Vacuum Bottle (autoclavable PP or chemically-resistant HDPE), each with a patented VersaCap closure for tangle-free openingno need to disconnect tubing, ever. A 0.2m PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nested neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for modulation of vacuum. Other Vactrap options offer one bottle, or setups without secondary containment. Say goodbye to glass. Say hello to Vactrap.