Learn More
Bellco Glass, Inc. Vactrap G, 1L + 1L, Glass Bottles, Red Bin, GL45 Cap w/1/4" ID Tubing, 1/EA
SDP
Catalog No. 502550720
Encompass_Preferred
VactrapG (Glass) is the ultimate vacuum trap system, adhering to the strictest CDC recommendations for laboratory safety. With its new continuous full vacuum rating, it is ideal for protecting vacuum lines and users. VactrapG safely contains waste produced during supernatant removal, chemical separation, and cell or tissue culture media aspiration.Following in the footsteps of previous innovations, VactrapG combines ease-of-use with revolutionary options and features to produce a new step forward in vacuum trap technology. From PUREGRIP glass media bottle versions to the inclusion of Foxx’s tangle-free VersaCap closures, to a brand new extra length tube, scientists now have access to better solutions at a lower cost.
Catalog No. 50-255-0720 Supplier Bellco Glass, Inc. Supplier No. 452800035
VactrapG (Glass) is the ultimate vacuum trap system, adhering to the strictest CDC recommendations for laboratory safety. With its new continuous full vacuum rating, it is ideal for protecting vacuum lines and users. VactrapG safely contains waste produced during supernatant removal, chemical separation, and cell or tissue culture media aspiration.Following in the footsteps of previous innovations, VactrapG combines ease-of-use with revolutionary options and features to produce a new step forward in vacuum trap technology. From PUREGRIP glass media bottle versions to the inclusion of Foxx’s tangle-free VersaCap closures, to a brand new extra length tube, scientists now have access to better solutions at a lower cost.