Learn More
Foxx Life Sciences Vactrap™ Secondary Container Spill Basin, Safety Tray with Dividers, for use with Vactrap™ Vacuum Trap System or for General Lab Safety, Waste Storage, and Spill Containment
Catalog No. 501489332
Encompass_Preferred
300-0005-FLS: Vactrap™ Red Bin w/ DividersFoxx Life Sciences Vactrap is the worlds most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap comes ready to usesimply attach to your vacuum and begin your work. The Vactrap Bin serves as a super-stable basin for secondary containment of the Vactrap system. Complete with dividers to hold bottles firmly in place, the bin vastly increases stability while maintaining a small footprint. Also great for use for general secondary containment outside of vacuum or aspiration applications.Say goodbye to glass. Say hello to Vactrap.
Catalog No. 50-148-9332 Supplier Foxx Life Sciences Supplier No. 3000005FLS
300-0005-FLS: Vactrap™ Red Bin w/ DividersFoxx Life Sciences Vactrap is the worlds most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap comes ready to usesimply attach to your vacuum and begin your work. The Vactrap Bin serves as a super-stable basin for secondary containment of the Vactrap system. Complete with dividers to hold bottles firmly in place, the bin vastly increases stability while maintaining a small footprint. Also great for use for general secondary containment outside of vacuum or aspiration applications.Say goodbye to glass. Say hello to Vactrap.