Learn More
Foxx Life Sciences Vactrap™ 2, XXL Secondary Container Spill Basin Safety Tray with Dividers, for use with Vactrap™ 2 Vacuum Trap System or for General Lab Safety, Waste Storage, and Spill Containment
Catalog No. 502060932
Encompass_Preferred
Foxx Life Sciences' Vactrap™ 2 is the world's most advanced vacuum trap system. Ideal for the protection of vacuum lines and scientists alike, Vactrap™ 2 safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap™ 2 comes ready to use; simply attach to your vacuum and begin your work. The Vactrap™ 2 Bin serves as a super-stable basin for secondary containment of the Vactrap™ 2 system. Complete with dividers to hold bottles firmly in place, the bin vastly increases stability while maintaining a small footprint. Also great for use for general secondary containment outside of vacuum or aspiration applications. This bin is autoclavable. Say goodbye to glass. Say hello to Vactrap™ 2.
Catalog No. 50-206-0932 Supplier Foxx Life Sciences Supplier No. 3000010FLS
Foxx Life Sciences' Vactrap™ 2 is the world's most advanced vacuum trap system. Ideal for the protection of vacuum lines and scientists alike, Vactrap™ 2 safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap™ 2 comes ready to use; simply attach to your vacuum and begin your work. The Vactrap™ 2 Bin serves as a super-stable basin for secondary containment of the Vactrap™ 2 system. Complete with dividers to hold bottles firmly in place, the bin vastly increases stability while maintaining a small footprint. Also great for use for general secondary containment outside of vacuum or aspiration applications. This bin is autoclavable. Say goodbye to glass. Say hello to Vactrap™ 2.