Learn More
Foxx Life Sciences Vactrap™ Tubing Extension, 4 feet (4'), with 0.2um PTFE Inline Vent Filter
Catalog No. 501489329
Encompass_Preferred
300-0001-FLS: Vactrap™ Tubing Extension, 4 ft., w/ 0.2um PTFE Vent FilterFoxx Life Sciences Vactrap is the worlds most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap comes ready to usesimply attach to your vacuum and begin your work. Vactrap tubing extensions allow users to replace or extend the tubing connecting their Vactrap system to their vacuum. Available in 4, 6 or 8-foot variations, these extensions improve Vactrap versatility and mobility in the lab. Each extension includes a replacement inline vent filter. For more replacement vent filters, search Foxx vent filter. Say goodbye to glass. Say hello to Vactrap.
Catalog No. 50-148-9329 Supplier Foxx Life Sciences Supplier No. 3000001FLS
300-0001-FLS: Vactrap™ Tubing Extension, 4 ft., w/ 0.2um PTFE Vent FilterFoxx Life Sciences Vactrap is the worlds most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap comes ready to usesimply attach to your vacuum and begin your work. Vactrap tubing extensions allow users to replace or extend the tubing connecting their Vactrap system to their vacuum. Available in 4, 6 or 8-foot variations, these extensions improve Vactrap versatility and mobility in the lab. Each extension includes a replacement inline vent filter. For more replacement vent filters, search Foxx vent filter. Say goodbye to glass. Say hello to Vactrap.