Learn More
Bellco Glass, Inc. Vactrap XL, Red Bin With Dividers, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800005
Encompass_Preferred
Vactrap XL is the world’s most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap XL safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap XL comes ready to use; simply attach to your vacuum and begin your work.The Vactrap XL Bin serves as a super-stable basin for secondary containment of the Vactrap XL system. Complete with dividers to hold bottles firmly in place, the bin vastly increases stability while maintaining a small footprint. Also great for use for general secondary containment outside of vacuum or aspiration applications. This bin is autoclaveable.Say goodbye to glass. Say hello to Vactrap XL.Specifications:Height: 5″ (127mm)Length: 16.5″ (419.1mm)Width: 10.87″ (276.2mm)Bin Material: PolypropyleneAutoclavable
Catalog No. 50-255-0750