Learn More
CHEMetrics™ by AquaPhoenix Vacu-vials Ammonia Water Quality Instrumental Test Kit, 0 to 10 ppm and 0 to 150 ppm
SDP
Catalog No. 13299567
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-567 30
1 options
Catalog No. 13-299-567 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K1513
Add to Cart
Add to Cart
Instrumental ammonia test kit using Direct Nesslerization and Hydroxybenzyl Alcohol methods. Designed for use with compatible photometers or spectrophotometers that accept a 13 mm round cell.
  • Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, Stabilizer Solution, 25 mL sample cup, 3-mL syringe, ampoule blank, and instructions.
  • Ammonia concentrations between 10 and 150 ppm can be determined using the 3-mL syringe and dilution instructions included.
Specifications
Analytes Ammonia
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 10.00 and 0 to 150 ppm
Type Water Quality Instrumental Test Kit