Learn More
CHEMetrics™ by AquaPhoenix Vacu-vials Ammonia Water Quality Instrumental Test Kit, HBA
SDP
Catalog No. 13299566
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-566 30
1 options
Catalog No. 13-299-566 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K1413
Add to Cart
Add to Cart
Instrumental ammonia test kit using Direct Nesslerization and Hydroxybenzyl Alcohol methods. Designed for use with compatible photometers or spectrophotometers that accept a 13 mm round cell.
  • This ammonia test kit comes in a cardboard box and contains everything needed to perform 30 tests (except distilled water): 30 ampoules, Stabilizer Solution, Catalyzer Solution, Activator Solution, 25 mL sample cup, 3 mL syringe, ampoule blank, and instructions.
  • This kit can be used to determine free ammonia concentration in the presence of monochloramine.
Specifications
Analytes Ammonia
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 3.00 and 0 to 60.0 ppm
Type Water Quality Instrumental Test Kit