Learn More
CHEMetrics™ by AquaPhoenix Vacu-vials Chloride Water Quality Instrumental Test Kit
SDP
Catalog No. 13299546
Change view
Click to view available options
No. of Tests:
29
Catalog No. No. of Tests
13-299-546 29
1 options
Catalog No. 13-299-546 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K2103
Add to Cart
Add to Cart
Instrumental chloride test kit supplied in a cardboard box with components for 30 tests. Requires a compatible photometer or spectrophotometer for analysis.
  • Kit K-2103 comes in a cardboard box and contains everything needed to perform up to 29 tests (except distilled water).
  • The kit includes thirty ampoules, Activator Solution, a 25 mL sample cup, ampoule blank, a 1.0 mL syringe, instructions, and MSDS.
Specifications
Analytes Chloride
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 29
Range 0 to 40.0 ppm
Type Water Quality Instrumental Test Kit